3-(Difluoromethoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A171652-5G A171652-100MG A171652-10G A171652-250MG A171652-25G
103806-650EA 55 USD
103806-650 103844-926 77082-088 77236-286 77236-288 77236-290 77301-142
3-(Difluoromethoxy)benzoic acid 97%
3-(Difluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MW: 188.13 g/mol
Boiling Pt: 287.3 °C (760 mm Hg)
Melting Pt: 105 °C
Flash Pt: 127.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236224
CAS Number: 4837-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR