3-(Difluoromethoxy)-4-fluorobenzaldehyde 95%

Supplier: Ambeed
Warning

A225082-100MG A225082-250MG
103819-154EA 131 USD
103819-154 77098-050 77098-052
3-(Difluoromethoxy)-4-fluorobenzaldehyde 95%
3-(Difluoromethoxy)-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD14698501
CAS Number: 1214367-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR