3-(Dibromomethyl)picolinonitrile 95%
Supplier: Ambeed
|
Warning
|
A100712-1G
A100712-5G
103795-588EA
43
USD
103795-588
103821-286
77296-824
77296-826
3-(Dibromomethyl)picolinonitrile 95%
3-(Dibromomethyl)picolinonitrile
Precaución: for Research Use Only
Formula:
C₇H₄Br₂N₂ MW: 275.93 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11040632 CAS Number: 126570-65-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...