3-(Chlorosulfonyl)benzoic acid 96%
Supplier: Ambeed
|
Danger
|
A439989-25G
A439989-5G
A439989-10G
A439989-1G
103827-652EA
7
USD
103827-652
103838-718
76842-474
76842-476
77143-370
77307-306
3-(Chlorosulfonyl)benzoic acid 96%
3-(Chlorosulphonyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₇H₅ClO₄S Boiling Pt: 402.5 °C (760 mm Hg) Melting Pt: 128…130 °C Flash Pt: 197.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024877 CAS Number: 4025-64-3 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...