3-(Chloromethyl)pyridazine hydrochloride 97%

Supplier: Ambeed
Danger

A167525-250MG A167525-1G A167525-5G
103806-198EA 251 USD
103806-198 103844-458 76872-256 77300-902 77300-904
3-(Chloromethyl)pyridazine hydrochloride 97%
3-(Chloromethyl)pyridazine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₅ClN₂·ClH
MW: 165.021 g/mol
Boiling Pt: 282.3 °C (760 mm Hg)
Flash Pt: 151.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19705404
CAS Number: 27349-66-2
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR