3-(Chloromethyl)benzonitrile 97%

Supplier: Ambeed
Danger

Synonyms: 3-Cyanobenzyl chloride

A127536-100G A127536-10G A127536-25G A127536-5G
77109-000EA 83.82 USD
77109-000 77109-002 77313-210 77313-212
3-(Chloromethyl)benzonitrile 97%
3-(Chloromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆ClN
MW: 151.5900 g/mol
Melting Pt: 66…71 °C
Flash Pt: 111.4 °C
Storage Temperature: Ambient
MDL Number: MFCD02684191
CAS Number: 64407-07-4
UN: 2923
ADR: 8,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR