3-(Chloromethyl)-2-methylpyridine hydrochloride 95%

Supplier: Ambeed
Warning

A212440-100MG A212440-250MG
103818-202EA 191 USD
103818-202 103848-898 76814-604 76814-606
3-(Chloromethyl)-2-methylpyridine hydrochloride 95%
3-(Chloromethyl)-2-methylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD09753485
CAS Number: 58539-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR