3-(Carboxymethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A226207-250MG A226207-1G A226207-5G
103819-232EA 52 USD
103819-232 103850-132 76815-042 77088-858 77088-860
3-(Carboxymethyl)benzoic acid 95%
3-(Carboxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
Boiling Pt: 412.8 °C (760 mm Hg)
Flash Pt: 217.6 °C
Storage Temperature: Ambient
MDL Number: MFCD15523567
CAS Number: 2084-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR