3-Picolyl bromide hydrobromide 96%

Supplier: Ambeed
Danger

Synonyms: 3-(Bromomethyl)pyridine hydrobromide

A568387-1G A568387-25G A568387-5G A568387-100G A568387-10G
103828-790EA 9 USD
103828-790 103839-932 76844-050 76844-052 76844-054 77103-664 77103-666
3-Picolyl bromide hydrobromide 96%
3-Picolyl bromide hydrobromide

Precaución: for Research Use Only

Formula: C₆H₇Br₂N
MW: 252.94 g/mol
Melting Pt: 150…155 °C
Flash Pt: 114.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01321314
CAS Number: 4916-55-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR