3-(Bromomethyl)phenyl acetate 95%

Supplier: Ambeed
Warning

A341741-1G A341741-100MG
103825-842EA 178 USD
103825-842 103836-816 76839-846 77015-596
3-(Bromomethyl)phenyl acetate 95%
3-(Bromomethyl)phenyl acetate

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.071 g/mol
Boiling Pt: 255.1±15.0 °C (760 mm Hg)
Flash Pt: 108.1 °C
Storage Temperature: Freezer
MDL Number: MFCD08543447
CAS Number: 49617-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR