3-(Bromomethyl)oxetane 95%

Supplier: Ambeed
Warning

A173623-100MG A173623-250MG A173623-1G
103806-848EA 88 USD
103806-848 103845-148 76985-556 76985-558 77082-408
3-(Bromomethyl)oxetane 95%
3-(Bromomethyl)oxetane

Precaución: for Research Use Only

Formula: C₄H₇BrO
MW: 151.002 g/mol
Storage Temperature: Freezer
MDL Number: MFCD20278240
CAS Number: 1374014-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR