3-(Bromomethyl)benzamide 95%

Supplier: Ambeed
Danger

A143879-100MG A143879-250MG
103803-336EA 190 USD
103803-336 76823-568 77885-208
3-(Bromomethyl)benzamide 95%
3-(Bromomethyl)benzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD23135264
CAS Number: 509073-67-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR