3-(Bromomethyl)benzaldehyde 97%

Supplier: Ambeed
Danger

A216229-1G A216229-10G A216229-250MG A216229-5G
103818-542EA 31 USD
103818-542 103849-254 77087-828 77275-316 77275-318 77303-428
3-(Bromomethyl)benzaldehyde 97%
3-(Bromomethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO
Boiling Pt: 269.7 °C (760 mm Hg)
Melting Pt: 49 °C
Flash Pt: 101.268 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797497
CAS Number: 82072-23-9
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR