3-Bromomethyl-3-methyloxetane 97%

Supplier: Ambeed
Danger

Synonyms: 2-(Bromomethyl)-2-methyl-1,3-epoxypropane, 3-(Bromomethyl)-3-methyloxetane

A658121-1G A658121-25G A658121-5G A658121-10G
103840-428EA 21 USD
103840-428 76844-618 76844-620 76844-622 77280-552
3-Bromomethyl-3-methyloxetane 97%
3-Bromomethyl-3-methyloxetane

Precaución: for Research Use Only

Formula: C₅H₉BrO
MW: 165.04 g/mol
Boiling Pt: 162 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02684290
CAS Number: 78385-26-9
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR