(+/-)-3-(Boc-aminomethyl)piperidine 95%

Supplier: Ambeed
Danger

Synonyms: 3-(Aminomethyl)piperidine, 3-BOC protected

A749377-100MG A749377-1G A749377-5G A749377-250MG
103840-842EA 35 USD
103840-842 76776-214 76845-070 76845-072 77627-496
(+/-)-3-(Boc-aminomethyl)piperidine 95%
(+/-)-3-(Boc-aminomethyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.309 g/mol
Melting Pt: 60…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01632454
CAS Number: 142643-29-6
UN: 3259
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR