3-(Bis(benzyloxy)phosphoryl)propanoic acid 95%

Supplier: Ambeed
Warning

A181500-100MG A181500-250MG
103807-612EA 258 USD
103807-612 77163-902 77163-904
3-(Bis(benzyloxy)phosphoryl)propanoic acid 95%
3-(Bis(benzyloxy)phosphoryl)propanoic acid

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₁₇H₁₉O₅P
MW: 334.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08064320
CAS Number: 805243-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR