3-(Benzyloxy)phenol 96%

Supplier: Ambeed
Warning

A117431-100G A117431-10G A117431-250MG A117431-25G A117431-5G
103798-888EA 22 USD
103798-888 103824-772 77108-334 77108-336 77108-338 77108-340 77311-556
3-(Benzyloxy)phenol 96%
3-(Benzyloxy)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₂
MW: 200.2332 g/mol
Boiling Pt: 358 °C (760 mm Hg)
Melting Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00134682
CAS Number: 3769-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR