3-(Benzyloxy)cyclobutanone 97%

Supplier: Ambeed
Warning

A107708-1G A107708-5G A107708-10G A107708-25G
103797-236EA 14 USD
103797-236 103823-004 77309-858 77309-860 77549-074 77592-154
3-(Benzyloxy)cyclobutanone 97%
3-(Benzyloxy)cyclobutanone

Precaución: for Research Use Only

Formula: C11H12O2
MW: 176.21 g/mol
Boiling Pt: 284.8 °C (760 mm Hg)
Flash Pt: 126 °C
Storage Temperature: Freezer
MDL Number: MFCD09755991
CAS Number: 30830-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR