3-(Benzyloxy)benzene-1,2-diamine 98%

Supplier: Ambeed
Warning

A371497-100MG A371497-250MG
103826-482EA 103 USD
103826-482 103837-500 77016-000 77016-002
3-(Benzyloxy)benzene-1,2-diamine 98%
3-(Benzyloxy)benzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD24646615
CAS Number: 89521-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR