2-Amino-3-benzyloxy-5-bromopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Benzyloxy)-5-bromopyridin-2-amine

A249349-250MG A249349-1G A249349-5G
103811-868EA 22 USD
103811-868 103851-706 77113-418 77304-480 77304-482
2-Amino-3-benzyloxy-5-bromopyridine 98%
2-Amino-3-benzyloxy-5-bromopyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁BrN₂O
MW: 279.13 g/mol
Boiling Pt: 376.7 °C (760 mm Hg)
Melting Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD11109946
CAS Number: 754230-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR