3-(Benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A156580-100G A156580-10G A156580-25G A156580-1G A156580-5G
103804-906EA 5 USD
103804-906 103843-106 77150-678 77150-680 77150-682 77318-898 77318-900
3-(Benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid 95%
3-(Benzyloxy)-4-oxo-4H-pyran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₅
MW: 246.21 g/mol
Boiling Pt: 487.5±45.0 °C (760 mm Hg)
Melting Pt: 150.1…151.8 °C
Flash Pt: 192.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22741607
CAS Number: 119736-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR