3-(3-Azetidinyl)pyridine dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Azetidin-3-yl)pyridine dihydrochloride

A135518-100MG A135518-250MG A135518-1G
103802-168EA 290 USD
103802-168 76823-204 76823-206 77315-000
3-(3-Azetidinyl)pyridine dihydrochloride 97%
3-(3-Azetidinyl)pyridine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀N₂·2ClH
MW: 207.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20527356
CAS Number: 1236791-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR