3-(Aminomethyl)pyridin-2(1H)-one hydrochloride 95%

Supplier: Ambeed
Warning

A149938-100MG A149938-250MG
103804-104EA 343 USD
103804-104 103842-232 77259-214 77259-216
3-(Aminomethyl)pyridin-2(1H)-one hydrochloride 95%
3-(Aminomethyl)pyridin-2(1H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉ClN₂O
MW: 160.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12035456
CAS Number: 85468-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR