[6-(Trifluoromethyl)pyridin-3-yl]methylamine 95%

Supplier: Ambeed
Danger

Synonyms: 3-(Aminomethyl)-6-(trifluoromethyl)pyridine

A225171-1G A225171-25G A225171-5G A225171-10G A225171-250MG
103819-162EA 90 USD
103819-162 103850-054 77088-728 77088-730 77088-732 77165-338 77165-340
[6-(Trifluoromethyl)pyridin-3-yl]methylamine 95%
[6-(Trifluoromethyl)pyridin-3-yl]methylamine

Precaución: for Research Use Only

Formula: C₇H₇F₃N₂·HCl
MW: 176.14 g/mol
Flash Pt: 79.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862649
CAS Number: 387350-39-2
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR