tert-Butyl-3-(aminomethyl)phenylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Aminomethyl)aniline, 1-BOC protected, 3-(Aminomethyl)-1-N-Boc-aniline

A374216-1G A374216-250MG A374216-10G
103826-546EA 19 USD
103826-546 103837-564 76840-840 77016-020 77594-120
tert-Butyl-3-(aminomethyl)phenylcarbamate 98%
tert-Butyl-3-(aminomethyl)phenylcarbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
Boiling Pt: 299.5 °C (760 mm Hg)
Melting Pt: 135…138 °C
Flash Pt: 134.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01317790
CAS Number: 205318-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR