3-(4-Phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ibrutinib deacryloylpiperidine
A150414-100G
A150414-10G
A150414-250MG
A150414-1G
A150414-25G
A150414-5G
103804-166EA
7
USD
103804-166
103842-300
77010-726
77010-728
77010-730
77317-814
77317-816
77317-818
3-(4-Phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine 98%
3-(4-Phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Precaución: for Research Use Only
Formula:
C₁₇H₁₃N₅O MW: 303.31 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20270360 CAS Number: 330786-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...