4-Nitrocinnamaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(4-Nitrophenyl)acrylaldehyde
A196918-250MG
A196918-5G
103816-876EA
18
USD
103816-876
103847-490
77287-578
77302-540
4-Nitrocinnamaldehyde 98%
4-Nitrocinnamaldehyde
Precaución: for Research Use Only
Formula:
C₉H₇NO₃ MW: 177.16 g/mol Boiling Pt: 311.6 °C (760 mm Hg) Melting Pt: 138…142 °C Flash Pt: 151.6 °C Storage Temperature: Freezer |
MDL Number:
MFCD00007379 CAS Number: 1734-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...