3-(4-Nitro-1-oxoisoindolin-2-yl)piperidine-2,6-dione 97%

Supplier: Ambeed
Warning

A427855-10G A427855-250MG A427855-1G A427855-5G
103827-490EA 19 USD
103827-490 103838-544 76829-504 76829-506 76842-204 76842-206
3-(4-Nitro-1-oxoisoindolin-2-yl)piperidine-2,6-dione 97%
3-(4-Nitro-1-oxoisoindolin-2-yl)piperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₁N₃O₅
Boiling Pt: 616.6 °C (760 mm Hg)
Flash Pt: 326.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11977267
CAS Number: 827026-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR