3-(4-Methylpiperazin-1-yl)aniline 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Aminophenyl)-4-methylpiperazine, 3-(4-Methyl-1-piperazinyl)aniline

A182656-10G A182656-250MG A182656-25G A182656-5G A182656-1G
103807-736EA 16 USD
103807-736 103846-112 76799-786 76799-788 77083-662 77083-664 77301-798
3-(4-Methylpiperazin-1-yl)aniline 98%
3-(4-Methylpiperazin-1-yl)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃
MW: 191.28 g/mol
Boiling Pt: 353 °C (760 mm Hg)
Melting Pt: 98…99 °C
Flash Pt: 164.9 °C
Storage Temperature: Ambient
MDL Number: MFCD08435857
CAS Number: 148546-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR