3-(4-Methoxyphenyl)pyridine 97%

Supplier: Ambeed
Warning

A403143-100MG A403143-250MG A403143-1G
103827-086EA 204 USD
103827-086 77278-770 77278-772 77307-084
3-(4-Methoxyphenyl)pyridine 97%
3-(4-Methoxyphenyl)pyridine

Precaución: Inert atmosphere, room temperature

Formula: C₁₂H₁₁NO
MW: 185.22 g/mol
Melting Pt: 62…63 °C at N/A mmHg
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD06801825
CAS Number: 5958-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR