3-(4-Methoxyphenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Methoxyphenyl)propanoic acid

A810857-100G A810857-10G A810857-500G A810857-25G A810857-5G
103841-092EA 6 USD
103841-092 76776-568 76776-570 76776-572 76845-344 76845-346
3-(4-Methoxyphenyl)propionic acid 98%
3-(4-Methoxyphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 318.5 °C (760 mm Hg)
Melting Pt: 99…103 °C
Flash Pt: 125.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00002777
CAS Number: 1929-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR