3-(4-Methoxyphenyl)propan-1-ol 95%

Supplier: Ambeed
Warning

A268311-1G A268311-25G A268311-5G A268311-100G A268311-10G
103812-912EA 25 USD
103812-912 103852-814 77092-566 77092-568 77092-570 77276-722 77276-724
3-(4-Methoxyphenyl)propan-1-ol 95%
3-(4-Methoxyphenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Boiling Pt: 283.9 °C (760 mm Hg)
Melting Pt: 26 °C at N/A mmHg
Flash Pt: 122.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00002952
CAS Number: 5406-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR