3-(4-Methoxyphenyl)-5-(piperidin-3-yl)-1,2,4-oxadiazole hydrochloride 98%

Supplier: Ambeed
Warning

A1003148-100MG A1003148-250MG
103795-362EA 629 USD
103795-362 76646-004 76646-006
3-(4-Methoxyphenyl)-5-(piperidin-3-yl)-1,2,4-oxadiazole hydrochloride 98%
3-(4-Methoxyphenyl)-5-(piperidin-3-yl)-1,2,4-oxadiazole hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₈ClN₃O₂
Storage Temperature: Refrigerator
CAS Number: 851882-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR