(+/-)-Equol 96%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Hydroxyphenyl)chroman-7-ol, (R,S)-Equol, 4′,7-Dihydroxyisoflavane, 7-Hydroxy-3-(4-hydroxyphenyl)chroman, (±)-Equol, 4',7-Isoflavandiol, 3-(4-Hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-7-ol

A168315-100G A168315-250MG A168315-1G
103806-274EA 11 USD
103806-274 103844-530 76872-284 76872-286 77081-586
(+/-)-Equol 96%
(+/-)-Equol

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
Melting Pt: 158…160 °C
Storage Temperature: Freezer
MDL Number: MFCD00016662
CAS Number: 94105-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR