3-(4-Hydroxyphenyl)-1-propanol 95%

Supplier: Ambeed
Warning

A174374-1G A174374-5G A174374-1MG A174374-250MG
103806-938EA 21 USD
103806-938 103845-236 77082-536 77082-538 77273-820 77273-822
3-(4-Hydroxyphenyl)-1-propanol 95%
3-(4-Hydroxyphenyl)-1-propanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
Boiling Pt: 311.6 °C (760 mm Hg)
Melting Pt: 51…54 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002953
CAS Number: 10210-17-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR