4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, NSC 2821, NSC 674320, NSC 51986, Coniferic acid, Ferulic acid

A137025-1000G A137025-100G A137025-100MG A137025-10G A137025-25MG A137025-500G A137025-25G
76688-840EA 165.51 USD
76688-840 76688-842 76688-844 76688-846 76688-848 76688-850 77315-328
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 98%
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
Melting Pt: 171 °C
Flash Pt: 150.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004400
CAS Number: 1135-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR