3-(4-Fluorophenyl)propionic acid 97%
Supplier: Ambeed
|
Warning
|
A241524-10G
A241524-250MG
A241524-1G
A241524-25G
A241524-5G
A241524-100G
103811-414EA
6
USD
103811-414
103851-218
76692-300
76692-302
77090-382
77090-384
77090-386
77626-284
3-(4-Fluorophenyl)propionic acid 97%
3-(4-Fluorophenyl)propionic acid
Precaución: for Research Use Only
Formula:
C₉H₉FO₂ MW: 168.17 g/mol Boiling Pt: 278.9 °C (760 mm Hg) Melting Pt: 86…91 °C Flash Pt: 122.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060327 CAS Number: 459-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...