4-Fluorobenzoylacetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Fluorophenyl)-3-oxopropanenitrile

A260205-100G A260205-10G A260205-500G A260205-1G A260205-25G A260205-5G
103812-486EA 5 USD
103812-486 103852-368 76692-682 76692-684 76692-686 77091-950 77091-952 77091-954
4-Fluorobenzoylacetonitrile 98%
4-Fluorobenzoylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₆FNO
MW: 163.15 g/mol
Boiling Pt: 311.9 °C (760 mm Hg)
Melting Pt: 84…88 °C
Flash Pt: 142.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00662062
CAS Number: 4640-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR