3-(4-Cyanophenyl)propanoic acid 97%
Supplier: Ambeed
|
Warning
|
A111219-10G
A111219-250MG
A111219-25G
A111219-1G
A111219-5G
103797-806EA
11
USD
103797-806
103823-620
77270-908
77270-910
77270-912
77310-422
77310-424
3-(4-Cyanophenyl)propanoic acid 97%
3-(4-Cyanophenyl)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₀H₉NO₂ Boiling Pt: 362.2 °C (760 mm Hg) Melting Pt: 136…140 °C Flash Pt: 172.881 °C Storage Temperature: Ambient |
MDL Number:
MFCD08234823 CAS Number: 42287-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...