3-(4'-Chlorophenyl)propanol 95%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Chlorophenyl)propan-1-ol

A160060-100G A160060-1G A160060-25G A160060-5G
103805-326EA 15 USD
103805-326 103843-556 77273-272 77300-436 77319-468 77319-470
3-(4'-Chlorophenyl)propanol 95%
3-(4'-Chlorophenyl)propanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
Boiling Pt: 270.7 °C (760 mm Hg)
Flash Pt: 117.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00831102
CAS Number: 6282-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR