3-(4-Bromophenyl)propan-1-ol 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Bromophenyl)propyl alcohol, 3-(4-Bromophenyl)-1-propanol

A120289-100G A120289-10G A120289-25G A120289-1G A120289-5G
103799-912EA 6 USD
103799-912 77271-456 77271-458 77271-460 77312-018 77312-020
3-(4-Bromophenyl)propan-1-ol 98%
3-(4-Bromophenyl)propan-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09028724
CAS Number: 25574-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR