4-Bromobenzoylacetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Bromophenyl)-3-oxopropanenitrile

A209546-1G A209546-5G A209546-100G A209546-10G A209546-250MG
103817-946EA 7 USD
103817-946 103848-632 77086-978 77086-980 77260-992 77260-994 77260-996
4-Bromobenzoylacetonitrile 98%
4-Bromobenzoylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.06 g/mol
Boiling Pt: 370 °C (760 mm Hg)
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00996063
CAS Number: 4592-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR