3-(4-Bromophenyl)-1H-[1,2,4]triazole 95%

Supplier: Ambeed
Warning

A148346-250MG A148346-1G A148346-5G
103803-920EA 48 USD
103803-920 103842-032 76972-960 77317-488 77317-490
3-(4-Bromophenyl)-1H-[1,2,4]triazole 95%
3-(4-Bromophenyl)-1H-[1,2,4]triazole

Precaución: for Research Use Only

Formula: C₈H₆BrN₃
MW: 224.0600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16659605
CAS Number: 118863-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR