3-(4-Bromobenzoyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Bromophenyl)-4-oxobutanoic acid

A113741-100G A113741-10G A113741-250MG A113741-25G A113741-500G A113741-1G A113741-5G
103798-286EA 11 USD
103798-286 103824-130 77271-072 77271-074 77271-076 77271-078 77271-080 77310-944 77310-946
3-(4-Bromobenzoyl)propionic acid 98%
3-(4-Bromobenzoyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₃
MW: 257.08 g/mol
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00016563
CAS Number: 6340-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR