3-Cyanophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 3-Cyanobenzeneboronic acid pinacol ester

A275177-100G A275177-25G A275177-5G A275177-1G A275177-10G
103813-240EA 10 USD
103813-240 103853-166 76723-136 77093-008 77093-010 77305-190 77601-216
3-Cyanophenylboronic acid pinacol ester 97%
3-Cyanophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
Boiling Pt: 341 °C (760 mm Hg)
Melting Pt: 78…82 °C
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD03789262
CAS Number: 214360-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR