3-Carboxyphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

A230904-100G A230904-10G A230904-250MG A230904-1G A230904-5G
103810-734EA 7 USD
103810-734 103850-480 76722-232 76722-234 76722-236 77089-352 77089-354
3-Carboxyphenylboronic acid pinacol ester 98%
3-Carboxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BO₄
Melting Pt: 206…211 °C
Flash Pt: 192.8 °C
Storage Temperature: Ambient
MDL Number: MFCD03411930
CAS Number: 269409-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR