3-Aminophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester

A223471-10G A223471-250MG A223471-5G A223471-1G A223471-100G
103819-040EA 7 USD
103819-040 103849-920 76722-026 76722-028 77088-542 77303-650 77600-792
3-Aminophenylboronic acid pinacol ester 97%
3-Aminophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₂
Boiling Pt: 351.2 °C (760 mm Hg)
Melting Pt: 89…94 °C
Flash Pt: 166.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03453668
CAS Number: 210907-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR