3-(4-tert-Butylphenyl)propionic acid 97%

Supplier: Ambeed
Warning

A122277-10G A122277-250MG A122277-1G A122277-5G
103800-198EA 18 USD
103800-198 77271-568 77271-570 77297-860 77297-862
3-(4-tert-Butylphenyl)propionic acid 97%
3-(4-tert-Butylphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
Boiling Pt: 318.6 °C (760 mm Hg)
Flash Pt: 215.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00796482
CAS Number: 1208-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR