3-(m-Tolyl)propionic acid 98%

Supplier: Ambeed
Warning

A839937-100MG A839937-10G A839937-250MG A839937-5G A839937-1G A839937-25G
103841-204EA 58 USD
103841-204 76698-310 76698-312 76698-314 76845-444 77308-434 77308-436
3-(m-Tolyl)propionic acid 98%
3-(m-Tolyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
Boiling Pt: 289.9 °C (760 mm Hg)
Melting Pt: 39…42 °C at N/A mmHg
Flash Pt: 187.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00060326
CAS Number: 3751-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR