3-(3-Hydroxyphenyl)propionic acid 98%
Supplier: Ambeed
|
Warning
|
A157385-10G
A157385-25G
A157385-5G
A157385-100G
A157385-250MG
103805-000EA
13
USD
103805-000
103843-202
77247-532
77300-258
77300-260
77746-077
77746-078
3-(3-Hydroxyphenyl)propionic acid 98%
3-(3-Hydroxyphenyl)propionic acid
Precaución: for Research Use Only
Formula:
C₉H₁₀O₃ Boiling Pt: 354.5 °C (760 mm Hg) Melting Pt: 111 °C Flash Pt: 182.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016554 CAS Number: 621-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...